2-Fluoro-5-methoxyphenylboronic acid

Supplier: Apollo Scientific
Warning

PC5056-1G PC5056-5G
APOSPC5056-1GEA 22.3 GBP
APOSPC5056-1G APOSPC5056-5G
2-Fluoro-5-methoxyphenylboronic acid
2-Fluoro-5-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MW: 169,9460 g/mol
Melting Pt: 194…196 °C
Storage Temperature: Ambient
MDL Number: MFCD04974115
CAS Number: 406482-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR