4-Fluoro-3-nitrobenzenesulphonamide

Supplier: Apollo Scientific

PC501637-1G PC501637-5G
APOSPC501637-1GEA 49.9 GBP
APOSPC501637-1G APOSPC501637-5G
4-Fluoro-3-nitrobenzenesulphonamide
4-Fluoro-3-nitrobenzenesulphonamide
Formula: C₆H₅FN₂O₄S
MW: 220,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08703185
CAS Number: 406233-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR