2-(Difluoromethyl)-1,3,5-trifluorobenzene

Supplier: Apollo Scientific
Warning

Synonyms: 2,4,6-Trifluorobenzal fluoride

PC49546-1G PC49546-250MG
APOSPC49546-1GEA 117 GBP
APOSPC49546-1G APOSPC49546-250MG
2-(Difluoromethyl)-1,3,5-trifluorobenzene
2-(Difluoromethyl)-1,3,5-trifluorobenzene
Formula: C₇H₃F₅
MW: 182,09 g/mol
Boiling Pt: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD14698643
CAS Number: 1214338-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR