1-Bromo-4-(difluoromethyl)benzene

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromobenzal fluoride

PC49528-100G PC49528-25G
APOSPC49528-100GEA 996 GBP
APOSPC49528-100G APOSPC49528-25G
1-Bromo-4-(difluoromethyl)benzene
1-Bromo-4-(difluoromethyl)benzene
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 100 °C/53mmHg
Density: 1.604 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD06657964
CAS Number: 51776-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR