Methyl-2-fluoro-6-methylbenzoate 98%

Supplier: Apollo Scientific

Synonyms: Methyl 2-fluoro-6-methylbenzoate

PC48750-1G PC48750-5G
APOSPC48750-1GEA 125 GBP
APOSPC48750-1G APOSPC48750-5G
Methyl-2-fluoro-6-methylbenzoate 98%
Methyl 2-fluoro-6-methylbenzoate
Formula: C₉H₉FO₂
MW: 168,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226310
CAS Number: 197516-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR