2-Chloro-5-(Trifluoromethyl)Pyrazine 97%

Supplier: Apollo Scientific
Danger

PC48362-1G PC48362-5G
APOSPC48362-1GEA 86.4 GBP
APOSPC48362-1G APOSPC48362-5G
2-Chloro-5-(Trifluoromethyl)Pyrazine 97%
2-Chloro-5-(trifluoromethyl)pyrazine
Formula: C₅H₂ClF₃N₂
MW: 182.53 g/mol
Boiling Pt: 85 °C/112mmHg
Storage Temperature: Ambient
MDL Number: MFCD13189478
CAS Number: 799557-87-2
UN: 3267
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR