5-Amino-2-(trifluoromethyl)pyridine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Amino-6-(trifluoromethyl)pyridine, 2-(Trifluoromethyl)pyridin-5-amine, 2-(Trifluoromethyl)-5-pyridylamine

PC4776-10G PC4776-1G PC4776-5G PC4776-25G
APOSPC4776-10GEA 80 GBP
APOSPC4776-10G APOSPC4776-1G APOSPC4776-5G APOSPC4776-25G
5-Amino-2-(trifluoromethyl)pyridine 98%
5-Amino-2-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162,11 g/mol
Melting Pt: 39…45 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01862656
CAS Number: 106877-33-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR