1-[(Ammoniooxy)methyl]-3-fluorobenzene chloride

Supplier: Apollo Scientific
Warning

Synonyms: O-(3-Fluorobenzyl)hydroxylamine hydrochloride

PC4626-1G PC4626-5G
APOSPC4626-1GEA 286 GBP
APOSPC4626-1G APOSPC4626-5G
1-[(Ammoniooxy)methyl]-3-fluorobenzene chloride
1-[(Ammoniooxy)methyl]-3-fluorobenzene chloride
Formula: C₇H₉ClFNO
Melting Pt: 218…225 °C
Storage Temperature: Ambient
MDL Number: MFCD01114585
CAS Number: 51572-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR