2-Amino-3'-fluoroacetophenone hydrochloride 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Fluorophenacylamine hydrochloride

PC4574-1G PC4574-250MG
APOSPC4574-1GEA 250 GBP
APOSPC4574-1G APOSPC4574-250MG
2-Amino-3'-fluoroacetophenone hydrochloride 98%
2-Amino-3'-fluoroacetophenone hydrochloride
Formula: C₈H₉ClFNO
Melting Pt: 201…204 °C
Storage Temperature: Ambient
MDL Number: MFCD08532459
CAS Number: 93102-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR