Heptafluoro-2,2,3-trichlorobutane 97%

Supplier: Apollo Scientific

PC4516-100G PC4516-25G
APOSPC4516-100GEA 197 GBP
APOSPC4516-100G APOSPC4516-25G
Heptafluoro-2,2,3-trichlorobutane 97%
Heptafluoro-2,2,3-trichlorobutane
Formula: C₄Cl₃F₇
Boiling Pt: 98 °C
Melting Pt: 4 °C
Density: 1,748 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00018065
CAS Number: 335-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR