5-Fluoro-1-(tetrahydro-2-furfuryl)uracil

Supplier: Apollo Scientific

Synonyms: Ftorafur, Tegafur, 5-Fluoro-1-(tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

PC450272-25G PC450272-5G
APOSPC450272-25GEA 66.9 GBP
APOSPC450272-25G APOSPC450272-5G
5-Fluoro-1-(tetrahydro-2-furfuryl)uracil
5-Fluoro-1-(tetrahydro-2-furfuryl)uracil
Formula: C₈H₉FN₂O₃
MW: 200,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012351
CAS Number: 17902-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR