N-[4-Bromo-3-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide

Supplier: Apollo Scientific

PC450085-1G PC450085-5G
APOSPC450085-1GEA 205 GBP
APOSPC450085-1G APOSPC450085-5G
N-[4-Bromo-3-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide
N-[4-Bromo-3-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide
Formula: C₉H₄BrF₆NO
Storage Temperature: Ambient
MDL Number: MFCD00607205
CAS Number: 156425-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR