Potassium thiophene-3-trifluoroborate

Supplier: Apollo Scientific

Synonyms: Potassium 3-thiophenetrifluoroborate, Potassium trifluoro(thiophen-3-yl)borate

PC4467-100G PC4467-25G
APOSPC4467-100GEA 523 GBP
APOSPC4467-100G APOSPC4467-25G
Potassium thiophene-3-trifluoroborate
Potassium thiophene-3-trifluoroborate
Formula: C₄H₃BF₃KS
MW: 190,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701626
CAS Number: 192863-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR