4-Bromo-3-(trifluoromethyl)benzamide 95+%

Supplier: Apollo Scientific

Synonyms: 4-Bromo-3-(Trifluoromethyl)benzamide

PC446192-1G PC446192-5G
APOSPC446192-1GEA 227 GBP
APOSPC446192-1G APOSPC446192-5G
4-Bromo-3-(trifluoromethyl)benzamide 95+%
4-Bromo-3-(trifluoromethyl)benzamide
Formula: C₈H₅BrF₃NO
MW: 268,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18089254
CAS Number: 1034690-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR