α-Methyl-4-(trifluoromethyl)benzylamine 98%

Supplier: Apollo Scientific
Danger

Synonyms: 1-(4-(Trifluoromethyl)phenyl)ethanamine

PC4461-1G PC4461-5G
APOSPC4461-1GEA 132 GBP
APOSPC4461-1G APOSPC4461-5G
α-Methyl-4-(trifluoromethyl)benzylamine 98%
alpha-Methyl-4-(trifluoromethyl)benzylamine
Formula: C₉H₁₀F₃N
MW: 189,18 g/mol
Boiling Pt: 76,5…77.5 °C/4.8mmHg
Density: 1,18 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD04038878
CAS Number: 15996-84-6
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR