Heptafluorobutyramide 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2,2,3,3,4,4,4-Heptafluorobutyramide, Perfluorobutyramide

PC4440-5G PC4440-10G
APOSPC4440-5GEA 19.7 GBP
APOSPC4440-5G APOSPC4440-10G
Heptafluorobutyramide 97%
Heptafluorobutyramide
Formula: C₄H₂F₇NO
MW: 213.05 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00039773
CAS Number: 662-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR