4-Fluoro-2-(trifluoromethyl)benzyl alcohol 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(4-Fluoro-2-(trifluoromethyl)phenyl)methanol
PC4375J-1G
PC4375J-5G
PC4375J-25G
APOSPC4375J-1GEA
19.7
GBP
APOSPC4375J-1G
APOSPC4375J-5G
APOSPC4375J-25G
4-Fluoro-2-(trifluoromethyl)benzyl alcohol 97%
4-Fluoro-2-(trifluoromethyl)benzyl alcohol
Formula:
C₈H₆F₄O MW: 194.13 g/mol Melting Pt: 55…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061170 CAS Number: 220227-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...