Methyl-4-fluoro-3-nitrobenzoate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 4-fluoro-3-nitrobenzoate, 4-Fluorobenzoic acid methyl ester

PC4359-100G PC4359-25G PC4359-50G
APOSPC4359-100GEA 70.9 GBP
APOSPC4359-100G APOSPC4359-25G APOSPC4359-50G
Methyl-4-fluoro-3-nitrobenzoate 98%
Methyl 4-fluoro-3-nitrobenzoate
Formula: C₈H₆FNO₄
MW: 199,14 g/mol
Boiling Pt: 109 °C/1mmHg
Melting Pt: 59…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00196161
CAS Number: 329-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR