3-(3-Fluoro-4-methylphenyl)-1,2,4-oxadiazole 98%
Supplier: Apollo Scientific
PC421209-100MG
PC421209-1G
PC421209-250MG
APOSPC421209-100MGEA
175
GBP
APOSPC421209-100MG
APOSPC421209-1G
APOSPC421209-250MG
3-(3-Fluoro-4-methylphenyl)-1,2,4-oxadiazole 98%
3-(3-Fluoro-4-methylphenyl)-1,2,4-oxadiazole
Formula:
C₉H₇FN₂O MW: 178.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13181200 CAS Number: 1146699-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...