4-Fluorophenyl methyl sulphone 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Fluoro-4-(methylsulphonyl)benzene, 1-Fluor-4-(methylsulphonyl)benzol
PC4174-25G
PC4174-5G
APOSPC4174-25GEA
57.6
GBP
APOSPC4174-25G
APOSPC4174-5G
4-Fluorophenyl methyl sulphone 98%
4-Fluorophenyl methyl sulfone
Formula:
C₇H₇FO₂S MW: 174.2 g/mol Melting Pt: 78…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039753 CAS Number: 455-15-2 EINECS: 207-237-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...