1-(Difluoromethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
Supplier: Apollo Scientific
|
Warning
|
PC412371-5G
PC412371-25G
APOSPC412371-5GEA
53.7
GBP
APOSPC412371-5G
APOSPC412371-25G
1-(Difluoromethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
1-(Difluoromethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formula:
C₁₀H₁₅BF₂N₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD17011852 CAS Number: 1206640-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...