2-Methyl-5-(trifluoromethyl)phenylboronic acid 95%

Supplier: Apollo Scientific
Warning

PC412114-1G PC412114-5G PC412114-250MG
APOSPC412114-1GEA 186 GBP
APOSPC412114-1G APOSPC412114-5G APOSPC412114-250MG
2-Methyl-5-(trifluoromethyl)phenylboronic acid 95%
2-Methyl-5-(trifluoromethyl)phenylboronic acid
Formula: C₈H₈BF₃O₂
MW: 203,9541 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11042432
CAS Number: 947533-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR