2,4-Difluoro-3-formylphenylboronic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: (2,4-Difluoro-3-formylphenyl)boronic acid

PC412092-25G PC412092-5G
APOSPC412092-25GEA 434 GBP
APOSPC412092-25G APOSPC412092-5G
2,4-Difluoro-3-formylphenylboronic acid 95%
2,4-Difluoro-3-formylphenylboronic acid
Formula: C₇H₅BF₂O₃
MW: 185,9200 g/mol
Melting Pt: 206…210 °C
Storage Temperature: Ambient
MDL Number: MFCD04974116
CAS Number: 870718-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR