2-Bromo-N-[3-(trifluoromethyl)phenyl]acetamide 98%

Supplier: Apollo Scientific

Synonyms: 2-Bromo-N-(3-(trifluoromethyl)phenyl)acetamide, 2-Bromo-3'-(trifluoromethyl)acetanilide

PC408238-250MG PC408238-1G
APOSPC408238-250MGEA 55.1 GBP
APOSPC408238-250MG APOSPC408238-1G
2-Bromo-N-[3-(trifluoromethyl)phenyl]acetamide 98%
2-Bromo-N-[3-(trifluoromethyl)phenyl]acetamide
Formula: C₉H₇BrF₃NO
MW: 282,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00859566
CAS Number: 25625-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR