(Heptafluoropropyl)trimethylsilane

Supplier: Apollo Scientific
Warning

Synonyms: Trimethyl(heptafluoropropyl)silane, Trimethyl(perfluoropropyl)silane

PC408177-5G PC408177-1G PC408177-250MG
APOSPC408177-5GEA 165 GBP
APOSPC408177-5G APOSPC408177-1G APOSPC408177-250MG
(Heptafluoropropyl)trimethylsilane
(Heptafluoropropyl)trimethylsilane
Formula: C₆H₉F₇Si
Storage Temperature: Refrigerator
MDL Number: MFCD00216712
CAS Number: 3834-42-2
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR