Triethyl(trifluoromethyl)silane 97%

Supplier: Apollo Scientific
Danger

Synonyms: (Triethylsilyl)trifluoromethane

PC408176-25G PC408176-5G
APOSPC408176-25GEA 571 GBP
APOSPC408176-25G APOSPC408176-5G
Triethyl(trifluoromethyl)silane 97%
Triethyl(trifluoromethyl)silane
Formula: C₇H₁₅F₃Si
MW: 184,28 g/mol
Boiling Pt: 58 °C/60mmHg
Density: 0,98 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00192520
CAS Number: 120120-26-5
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR