4-Cyano-2,3,5,6-tetrafluorobenzotrifluoride

Supplier: Apollo Scientific
Warning

Synonyms: 2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzonitrile

PC408124-5G PC408124-10G
APOSPC408124-5GEA 197 GBP
APOSPC408124-5G APOSPC408124-10G
4-Cyano-2,3,5,6-tetrafluorobenzotrifluoride
4-Cyano-2,3,5,6-tetrafluorobenzotrifluoride
Formula: C₈F₇N
Storage Temperature: Ambient
MDL Number: MFCD04972776
CAS Number: 15895-68-8
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR