3,5-Dichloro-4-fluorobenzonitrile
Supplier: Apollo Scientific
|
Warning
|
PC408068-1G
PC408068-250MG
APOSPC408068-1GEA
55.1
GBP
APOSPC408068-1G
APOSPC408068-250MG
3,5-Dichloro-4-fluorobenzonitrile
3,5-Dichloro-4-fluorobenzonitrile
Formula:
C₇H₂Cl₂FN MW: 190,0019 g/mol Melting Pt: 52…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD09835204 CAS Number: 103879-31-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...