2-(4-Fluorophenyl)thiophene 95%

Supplier: Apollo Scientific

PC408001-100G PC408001-25G
APOSPC408001-100GEA 104 GBP
APOSPC408001-100G APOSPC408001-25G
2-(4-Fluorophenyl)thiophene 95%
2-(4-Fluorophenyl)thiophene
Formula: C₁₀H₇FS
MW: 178,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06802535
CAS Number: 58861-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR