4-Fluoro-2-nitrotoluene 98%

Supplier: Apollo Scientific
Warning

PC4040-100G PC4040-25G PC4040-500G
APOSPC4040-100GEA 22.3 GBP
APOSPC4040-100G APOSPC4040-25G APOSPC4040-500G
4-Fluoro-2-nitrotoluene 98%
4-Fluoro-2-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 137…139 °C/83mmHg
Melting Pt: 25…27 °C
Density: 1.26 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00007200
CAS Number: 446-10-6
EINECS: 207-161-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR