Ethyl-2-(trifluoromethyl)thiazole-5-carboxylate 97%

Supplier: Apollo Scientific
Warning

Synonyms: ethyl 2-(trifluoromethyl)-1,3-thiazole-5-carboxylate, Ethyl 2-(trifluoromethyl)thiazole-5-carboxylate

PC400725-25G PC400725-5G
APOSPC400725-25GEA 260 GBP
APOSPC400725-25G APOSPC400725-5G
Ethyl-2-(trifluoromethyl)thiazole-5-carboxylate 97%
Ethyl-2-(trifluoromethyl)thiazole-5-carboxylate
Formula: C₇H₆F₃NO₂S
MW: 225,1883 g/mol
Boiling Pt: 195 °C
Density: 1,34 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD09055619
CAS Number: 131748-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR