4-Bromo-3-(trifluoromethyl)-1H-pyrazole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Bromo-3-(trifluoromethyl)-1H-pyrazole
PC3672-1G
PC3672-5G
PC3672-25G
APOSPC3672-1GEA
19.7
GBP
APOSPC3672-1G
APOSPC3672-5G
APOSPC3672-25G
4-Bromo-3-(trifluoromethyl)-1H-pyrazole
4-Bromo-3-(trifluoromethyl)-1H-pyrazole
Formula:
C₄H₂BrF₃N₂ MW: 214.97 g/mol Melting Pt: 112…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD04969230 CAS Number: 19968-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...