2-Fluoro-4-methyl-5-pyridineboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluoro-4-methylpyridine-5-boronic acid

PC3614-1G PC3614-5G
APOSPC3614-1GEA 52.5 GBP
APOSPC3614-1G APOSPC3614-5G
2-Fluoro-4-methyl-5-pyridineboronic acid
2-Fluoro-4-methyl-5-pyridineboronic acid
Formula: C₆H₇BFNO₂
MW: 154,9400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08741406
CAS Number: 1072944-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR