3-Methyl-5-(trifluoromethoxy)benzaldehyde

Supplier: Apollo Scientific
Warning

PC3602-5G PC3602-1G PC3602-250MG
APOSPC3602-5GEA 154 GBP
APOSPC3602-5G APOSPC3602-1G APOSPC3602-250MG
3-Methyl-5-(trifluoromethoxy)benzaldehyde
3-Methyl-5-(trifluoromethoxy)benzaldehyde
Formula: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD08741401
CAS Number: 1000339-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR