(R)-(-)-3-Fluoropyrrolidine hydrochloride

Supplier: Apollo Scientific
Warning

Synonyms: (R)-(-)-3-Fluoropyrrolidine Hydrochloride

PC3455-1G PC3455-5G PC3455-10G
APOSPC3455-1GEA 95.7 GBP
APOSPC3455-1G APOSPC3455-5G APOSPC3455-10G
(R)-(-)-3-Fluoropyrrolidine hydrochloride
(R)-(-)-3-Fluoropyrrolidine hydrochloride
Formula: C₄H₉ClFN
MW: 125,57 g/mol
Melting Pt: 179…186 °C
Storage Temperature: Ambient
MDL Number: MFCD04038717
CAS Number: 136725-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR