2-Bromo-2',3'-difluoro-4'-(trifluoromethyl)acetophenone

Supplier: Apollo Scientific
Danger

Synonyms: 2,3-Difluoro-4-(trifluoromethyl)phenacyl bromide

PC3405-1G PC3405-5G
APOSPC3405-1GEA 89.2 GBP
APOSPC3405-1G APOSPC3405-5G
2-Bromo-2',3'-difluoro-4'-(trifluoromethyl)acetophenone
2-Bromo-2',3'-difluoro-4'-(trifluoromethyl)acetophenone
Formula: C₉H₄BrF₅O
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD09259004
CAS Number: 886762-02-3
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR