2-Amino-3-fluoropropanoic acid 97+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
H-β-Fluoro-DL-Ala-OH
PC3379-250MG
PC3379-100MG
APOSPC3379-250MGEA
905
GBP
APOSPC3379-250MG
APOSPC3379-100MG
2-Amino-3-fluoropropanoic acid 97+%
2-Amino-3-fluoropropanoic acid
Formula:
C₃H₆FNO₂ MW: 107,084 g/mol Melting Pt: 168 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00077524 CAS Number: 16652-37-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...