4,6-Difluoropyrimidine 95%
Supplier: Apollo Scientific
|
Danger
|
PC3333-25G
PC3333-5G
APOSPC3333-25GEA
354
GBP
APOSPC3333-25G
APOSPC3333-5G
4,6-Difluoropyrimidine 95%
Formula:
C₄H₂F₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD03412223 CAS Number: 2802-62-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...