2'-Fluoroacetanilide 98%
Supplier: Apollo Scientific
|
Warning
|
PC3320-100G
PC3320-25G
APOSPC3320-100GEA
122
GBP
APOSPC3320-100G
APOSPC3320-25G
2'-Fluoroacetanilide 98%
2'-Fluoroacetanilide
Formula:
C₈H₈FNO MW: 153,16 g/mol Boiling Pt: 140…142 °C/14mmHg Melting Pt: 76…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000315 CAS Number: 399-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...