Ethyl pentafluoropropionate 98%

Supplier: Apollo Scientific
Danger

PC3250-25G PC3250-10G PC3250-100G PC3250-1G PC3250-5G
APOSPC3250-25GEA 59.1 GBP
APOSPC3250-25G APOSPC3250-10G APOSPC3250-100G APOSPC3250-1G APOSPC3250-5G
Ethyl pentafluoropropionate 98%
Ethyl pentafluoropropionate

Undergoes Wittig reaction to give 1-perfluoroalkylenol ethers, which are useful precursors to other fluorine containing molecules: JOC., 57, 3807 (1992).

Formula: C₅H₅F₅O₂
MW: 192,09 g/mol
Boiling Pt: 73…76 °C
Density: 1,299 g/cm³ (25 °C)
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00000431
CAS Number: 426-65-3
EINECS: 207-043-6
UN: 3272
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR