1-(2-Fluoro-5-nitrophenyl)-1H-pyrrole

Supplier: Apollo Scientific

PC32128-1G PC32128-250MG PC32128-500MG PC32128-5G
APOSPC32128-1GEA 318 GBP
APOSPC32128-1G APOSPC32128-250MG APOSPC32128-500MG APOSPC32128-5G
1-(2-Fluoro-5-nitrophenyl)-1H-pyrrole
1-(2-Fluoro-5-nitrophenyl)-1H-pyrrole
Formula: C₁₀H₇FN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02180646
CAS Number: 96623-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR