Ethyl dibromofluoroacetate 97%
Supplier: Apollo Scientific
|
Danger
|
PC3185-100G
PC3185-25G
APOSPC3185-100GEA
530
GBP
APOSPC3185-100G
APOSPC3185-25G
Ethyl dibromofluoroacetate 97%
Ethyl dibromofluoroacetate
Formula:
C₄H₅Br₂FO₂ Boiling Pt: 82 °C/33mmHg Density: 1,894 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042068 CAS Number: 565-53-7 UN: 2922 ADR: 8+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...