Ethyl chlorofluoroacetate 97%
Supplier: Apollo Scientific
|
Danger
|
PC3180-100G
PC3180-25G
APOSPC3180-100GEA
217
GBP
APOSPC3180-100G
APOSPC3180-25G
Ethyl chlorofluoroacetate 97%
Formula:
C₄H₆ClFO₂ Boiling Pt: 128…130 °C Density: 1,225 g/cm³ (25 °C) Flash Pt: 44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039333 CAS Number: 401-56-9 EINECS: 206-930-5 UN: 2920 ADR: 8+3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...