1-Methyl-3-(trifluoromethyl)-1H-pyrazole

Supplier: Apollo Scientific
Warning

PC31708-10G PC31708-1G PC31708-5G
APOSPC31708-10GEA 840 GBP
APOSPC31708-10G APOSPC31708-1G APOSPC31708-5G
1-Methyl-3-(trifluoromethyl)-1H-pyrazole
1-Methyl-3-(trifluoromethyl)-1H-pyrazole
Formula: C₅H₅F₃N₂
MW: 150,104 g/mol
Boiling Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00179867
CAS Number: 154471-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR