2,6-Bis(trifluoromethyl)benzonitrile 98%

Supplier: Apollo Scientific
Danger

PC3096-1G PC3096-250MG PC3096-5G
APOSPC3096-1GEA 150 GBP
APOSPC3096-1G APOSPC3096-250MG APOSPC3096-5G
2,6-Bis(trifluoromethyl)benzonitrile 98%
2,6-Bis(trifluoromethyl)benzonitrile
Formula: C₉H₃F₆N
MW: 239,1172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061226
CAS Number: 25753-25-7
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR