2-Bromo-2',6'-difluoroacetophenone 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Bromo-1-(2,6-difluorophenyl)ethanone, 2,6-Difluorophenacyl bromide
PC3052-1G
PC3052-25G
PC3052-5G
APOSPC3052-1GEA
52.5
GBP
APOSPC3052-1G
APOSPC3052-25G
APOSPC3052-5G
2-Bromo-2',6'-difluoroacetophenone 98%
2-Bromo-2',6'-difluoroacetophenone
Formula:
C₈H₅BrF₂O MW: 235,0255 g/mol Boiling Pt: 88…97 °C/2mmHg Flash Pt: 96 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04038255 CAS Number: 56159-89-8 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...