2,3-Difluoro-4-pentylanisole 98%

Supplier: Apollo Scientific

PC303139-1G PC303139-5G
APOSPC303139-1GEA 285 GBP
APOSPC303139-1G APOSPC303139-5G
2,3-Difluoro-4-pentylanisole 98%
2,3-Difluoro-4-pentylanisole
Formula: C₁₂H₁₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD09258680
CAS Number: 134364-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR