4-Fluoro-2-(trifluoromethoxy)benzoic acid 97%
Supplier: Apollo Scientific
|
Warning
|
PC303136-5G
PC303136-1G
APOSPC303136-5GEA
603
GBP
APOSPC303136-5G
APOSPC303136-1G
4-Fluoro-2-(trifluoromethoxy)benzoic acid 97%
4-Fluoro-2-(trifluoromethoxy)benzoic acid
Formula:
C₈H₄F₄O₃ MW: 224,11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687173 CAS Number: 1242250-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...