4-Ethoxy-3,5-difluorobenzyl alcohol 98%

Supplier: Apollo Scientific

PC303086-10G PC303086-5G
APOSPC303086-10GEA 367 GBP
APOSPC303086-10G APOSPC303086-5G
4-Ethoxy-3,5-difluorobenzyl alcohol 98%
4-Ethoxy-3,5-difluorobenzyl alcohol
Formula: C₉H₁₀F₂O₂
MW: 188,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258690
CAS Number: 1017779-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR