4'-Chloro-3'-fluoropropiophenone 98%

Supplier: Apollo Scientific
Warning

PC302781-5G PC302781-1G
APOSPC302781-5GEA 133 GBP
APOSPC302781-5G APOSPC302781-1G
4'-Chloro-3'-fluoropropiophenone 98%
4'-Chloro-3'-fluoropropiophenone
Formula: C₉H₈ClFO
MW: 186,61 g/mol
Melting Pt: 30…34 °C
Storage Temperature: Ambient
MDL Number: MFCD09832382
CAS Number: 1017779-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR